Test kits for water testing, AQUAfast™ Powder Packs, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AC4P12 AC4P29 AC4P42 AC4P46 AC4P55 AC4P60 AC4P71 AC4P72 AC4P78 AC4P79 AC4P82 AC4P95 AC4P54
ORIOAC4P12EA 474 EUR
ORIOAC4P12 ORIOAC4P29 ORIOAC4P42 ORIOAC4P46 ORIOAC4P55 ORIOAC4P60 ORIOAC4P71 ORIOAC4P72 ORIOAC4P78 ORIOAC4P79 ORIOAC4P82 ORIOAC4P95 ORIOAC4P54
Test kits for water testing, AQUAfast™ Powder Packs, Orion™
Milieutestkits
Orion™ AQUAfast Powder Pack reagents perform colorimetric measurements of common water parameters with powders that dissolve quickly and mix easily.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast Colorimeters
Orion™ AQUAfast Powder Pack Reagents are designed for use with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Powder pack chemistries dissolve quickly in solution, are easy to mix, and do not require a stir rod or crushing. Each powder pack is notched for simplified opening in the lab or field.