Propagyl bromide ≥80% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
3-Bromopropyne
Stabilisatie:
Stabiised with magnesium oxide (MgO)
A16580.AD
A16580.AK
A16580.AU
A16580.ADEA
48.6
EUR
A16580.AD
A16580.AK
A16580.AU
Propagyl bromide ≥80% stabilised
Propagyl bromide
CAS No.: 106-96-7
Molecular Formula: HC≡CCH2Br
Formula Weight: 118.97
Storage Temperature: 2°C to 8°C
Flash Point: 3°(37°F)
Refractive Index: 1.4940
Melting Point: -61°
Boiling Point: 88-90°
Beilstein Reference: 605309
UN No.: 2924
MDL No.: MFCD00000241
Formule:
C₃H₃Br Kookpunt: 88…90 °C Smeltpunt: –61 °C Dichtheid: 1,335 Vlampunt: 3 °C (37 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00000241 CAS nummer: 106-96-7 EINECS: 203-447-1 UN: 2924 ADR: 3,II |