4-Bromo-2,3-dimethyl-6-nitroaniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB06740DA BTB06740EA BTB06740.50
MAYBBTB06740DAEA 0 EUR
MAYBBTB06740DA MAYBBTB06740EA MAYBBTB06740.50
4-Bromo-2,3-dimethyl-6-nitroaniline
4-Bromo-2,3-dimethyl-6-nitroaniline
Formule: C₈H₉BrN₂O₂
MW: 245.08 g/mol
MDL Nummer: MFCD00051710
CAS nummer: 108485-13-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR