n-Pentaan 98
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Pentane
17381.AK
17381.K2
17381.AKEA
30
EUR
17381.AK
17381.K2
n-Pentaan 98
n-Pentaan
Purity: 98%
CAS No.: 109-66-0
Molecular Formula: C5H12
Formula Weight: 72.15
Flash Point: -49°(-57°F)
Physical Form: Liquid
Density: 0.626
Refractive Index: 1.3580
Appearance: Colourless
Odour: Nearly Odourless
Melting Point: -130°
Boiling Point: 36°
Merck Reference: 14,7116
UN No.: 1265
MDL No.: MFCD00009498
Formule:
H₃C(CH₂)₃CH₃ MW: 72.15 g/mol Kookpunt: 36 °C Smeltpunt: –130 °C Dichtheid: 0.626 Vlampunt: –49 °C (–57 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009498 CAS nummer: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3 II Bereik: 01-2119459286-30 Merck Index: 13,07193 |