n-Pentaan ∼ 95%, EMPLURA®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Pentane
M-Clarity™ quality level = MQ200
1.07176.1000 1.07176.9190 1.07176.2500
1.07176.1000EA 71.9 EUR
1.07176.1000 1.07176.9190 1.07176.2500
n-Pentaan ∼ 95%, EMPLURA®, Supelco®
n-Pentaan
Formule: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kookpunt: 35…36 °C (1013 hPa)
Smeltpunt: –130 °C
Dichtheid: 0.63 g/cm³ (20 °C)
Vlampunt: –40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Bereik: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) ≥ 95.0 %
Identity (IR) conforms
Free acid (as CH₃COOH) ≤ 0.001 %
Density (d 20 °C/ 4 °C) 0.626 - 0.628
Readily carbonizable substances conforms
Evaporation residue ≤ 0.005 %