n-Undecaan ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Undecane
31807.18
31807.30
31807.18EA
52.5
EUR
31807.18
31807.30
n-Undecaan ≥99%
n-Undecaan
Purity: 99%
CAS No.: 1120-21-4
Molecular Formula: C11H24
Formula Weight: 156.30
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 0.74
Refractive Index: 1.4173
Appearance: Colourless
Odour: Mild
Melting Point: -26°
Boiling Point: 196°
UN No.: 2330
MDL No.: MFCD00008959
Formule:
H₃C(CH₂)₉CH₃ MW: 156.31 g/mol Kookpunt: 196 °C Smeltpunt: –26 °C Dichtheid: 0.74 Vlampunt: 60 °C (140 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008959 CAS nummer: 1120-21-4 EINECS: 214-300-6 UN: 2330 ADR: 3 III |