Sleeves for ground joint cones
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01101714 01101719 01101724 01101729 01101745
LENZ01101714EA 25.8 EUR
LENZ01101714 LENZ01101719 LENZ01101724 LENZ01101729 LENZ01101745
Sleeves for ground joint cones
Geslepen Geslepen hulzen
Sleeves, made of PTFE. For fitting onto standard ground cones.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from −200 to +280 °C
Sturdy construction with protruding grip so sleeves can be detached easily. With sealing rings on the outside of the ground socket, which prevent ground joints from seizing and give a very good seal. Grease-free connection; no grease residue to contaminate sample.