Serological pipettes
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1000833 1000841 1000854 1000860 1000870
612-1065EA 11 EUR
612-1065 HIRS1000841 612-1122 612-1123 HIRS1000870
Serological pipettes
Pipetten Serologische pipetten
Measuring pipettes made of AR-Glas®, class B.
  • With silver diffusion ink durable brown graduation
  • Wide opening, with expanded tip opening
  • Zero point at top, graduated to tip
  • With standard cotton plug (pipettes with 5 ml and more)