Graduated pipettes, type 3, Class AS, total delivery
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1130133 1130140 1130141
HIRS1130133EA 7.8 EUR
HIRS1130133 HIRS1130140 HIRS1130141
Graduated pipettes, type 3, Class AS, total delivery
Pipetten Gegradueerde pipetten
AR-Glas®.
  • Calibrated to deliver 'Ex'
  • With mouth piece for cotton plug, main point ring graduation
  • Graduated to tip, zero on top, type 3
  • Silver diffusion ink durable brown graduation
  • With dated batch identification
DIN EN ISO 835
Certificaten: DIN EN ISO 835. Conformity certified (DE-M marking).