Dophellingshoeken
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43058 43070 43078 43055 43066 43088
BRND43058EA 65.6 EUR
BRND43058 BRND43070 BRND43078 BRND43055 BRND43066 BRND43088
Dophellingshoeken
Flesdispenseropzet
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
  • Calibrated to deliver (TD, Ex)
  • Excellent chemical resistance
  • Excellent good temperature resistance
Automatic dispenser particularly suited for dispensing suspensions.
Bestelinformatie: Reservoir bottles available on request.