Slangaansluitingen, T-vormig
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01600508 01600510
229-3522EA 3.9 EUR
229-3522 LENZ01600510
Slangaansluitingen, T-vormig
Fittingen en connectoren
Tubing connectors made of DURAN® borosilicate glass,T-shaped with three glass olives.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • With tapered endpieces to ensure that tubes are held securely