Stopcocks for burettes, lateral
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02702001 02702501
LENZ02702001EA 27.1 EUR
LENZ02702001 LENZ02702501
Stopcocks for burettes, lateral
Afsluitkranen
NS stopcock made of borosilicate glass, facetted tip, with plug of solid glass or PTFE and screwthread retaining device.
  • Excellent chemical resistance
  • Excellent temperature resistance