Kolven, peervormig met twee halzen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
03222128 03221122 03221128 03221137 03221149 03222122 03222137 03222149
LENZ03222128EA 43.6 EUR
LENZ03222128 271-1232 271-1233 271-1234 LENZ03221149 LENZ03222122 LENZ03222137 LENZ03222149
Kolven, peervormig met twee halzen
Kolven Peervormige kolven
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.