Multiple adapters
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05005111 05005444
271-2221EA 37.7 EUR
271-2221 201-2217
Multiple adapters
Onderdelen voor destillatie Destillatieadapters met meerdere halzen
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.