Allyl bromide ≥99% stabilised
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
3-Bromopropene, 3-Propenyl bromide
Stabilisatie:
Stabilised with propylene oxide
Stabilized with 300-1000 ppm Propylene oxide
Purity: 99%
CAS No.: 106-95-6
Molecular Formula: C3H5Br
Formula Weight: 120.98
Flash Point: -2°(28°F)
Physical Form: Liquid
Density: 1.398
Refractive Index: 1.4700
Sensitivity: Light Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: -119°
Boiling Point: 70-71°
Merck Reference: 14,288
UN No.: 1099
MDL No.: MFCD00000244
Formule:
C₃H₅Br Kookpunt: 70…71 °C Smeltpunt: –119 °C Dichtheid: 1,398 Vlampunt: –2 °C (28 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000244 CAS nummer: 106-95-6 EINECS: 203-446-6 UN: 1099 ADR: 3,I Merck Index: 14,288 |