Allyl acetate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Acetic acid allyl ester
180651000 180655000
ACRO180651000EA 28.7 EUR
ACRO180651000 ACRO180655000
Allyl acetate 99%
Allyl acetate
Formule: C₅H₈O₂
MW: 100.12 g/mol
Kookpunt: 103
Smeltpunt: –96 °C
Dichtheid: 0.92 g/cm³
Vlampunt: 6
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008721
CAS nummer: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3 II
Merck Index: 12,00293
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %