Adapters met slangaansluiting
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05101004 05103001 05103003 05103002 05103004 05101001 05101002 05101003
LENZ05101004EA 16.9 EUR
LENZ05101004 LENZ05103001 271-2105 217-9092 LENZ05103004 LENZ05101001 LENZ05101002 LENZ05101003
Adapters met slangaansluiting
Adapters Adapters, Slijpstukken met slangaansluitingen
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Adapters with NS socket and straight glass olive.