Graduated pipettes, type 2, Class AS, total delivery
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1101341 1101335 1101333 1101329 1101353 1101354 1101342 1101370 1101365 1101360
612-4711EA 6.9 EUR
612-4711 612-4154 612-4153 HIRS1101329 612-4713 612-4156 612-4155 612-4159 612-4158 612-4157
Graduated pipettes, type 2, Class AS, total delivery
Pipetten Gegradueerde pipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex); Type 2: nominal volume on top
  • Marks and inscriptions in high contrast blue enamel
  • With standard cotton plug from five ml
  • Certificate of conformity with dated batch identification available on internet (www.hirschmannlab.de)
DIN EN ISO 835
Length: 360 mm ±5 mm except for 25 pipettes: 450 mm ±5 mm
Certificaten: DIN EN ISO 835. Conformity certified (DE-M marking).