Adapters met NS-afsluitkraan
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05104001 05104003 05104004 05105001 05105003 05105004 05105002
LENZ05104001EA 37.1 EUR
LENZ05104001 201-2107 LENZ05104004 LENZ05105001 LENZ05105003 LENZ05105004 LENZ05105002
Adapters met NS-afsluitkraan
Adapters Adapters, Slijpstukken met slangaansluitingen
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
With standard ground socket, standard ground stopcock with solid glass key, bore size 2,5 mm and a straight or right-angled tubing connector. The tubing connector has an external diameter of 8 mm.