Flenzen, KF, met glazen olijf
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05106216 05106110 05106116 05107110 05107116 05107216
LENZ05106216EA 64.5 EUR
LENZ05106216 LENZ05106110 LENZ05106116 LENZ05107110 LENZ05107116 LENZ05107216
Flenzen, KF, met glazen olijf
Adapters Adapters met kogelgewrichten
Made of DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes