Prefiltration capsules, Sartopure® GF Plus
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sartopure®
SARP5555305P9-OO-AEA 488 EUR
SARP5555305P9-OO-A SARP5555303P8-SS-A SARP5555305P0-SS-V
Prefiltration capsules, Sartopure® GF Plus
Filters Membraanfiltercapsules
Glass fibre, PP support layers. They are self-contained, ready to use filter units for removal of contaminants like colloids, lipids, protein aggregates and particles from bio-pharmaceutical fluids. They are used for protection of membrane filters, chromatography- and ultrafiltration systems.
  • Feature with highly charged glass fiber layers for effective clarification of fluid streams
  • Adsorptive depth filters assures outstanding total throughput performance