Burettes, straight stopcock, brown
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3150160 3150170 3150175 3150260 3150270 3150275
HIRS3150160EA 121 EUR
HIRS3150160 HIRS3150170 HIRS3150175 HIRS3150260 HIRS3150270 HIRS3150275
Burettes, straight stopcock, brown
Buretten
Burette, class AS, DURAN®, amber glass with white graduation.
  • Straight glass stopcock with glass key or valve stopcock with PTFE spindle
  • Main point ring graduation
  • Amber coloured for light sensitive fluids
  • Calibrated for EX
Certificaten: DIN EN ISO 385