Recirculating coolers, SemiChill series
Leverancier: JULABO GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
950005007P3H0 950005107P3H0 950010107P3H0 950002603P3H0 950002503P3H0
JULA950005007P3H0EA 16850 EUR
JULA950005007P3H0 JULA950005107P3H0 JULA950010107P3H0 JULA950002603P3H0 JULA950002503P3H0
Recirculating coolers, SemiChill series
Circulatiethermostaten Gesloten circulatiesystemen Koelingscirculatoren
Recirculating coolers can handle virtually any cooling requirements in laboratories or industrial environments efficiently.
  • Environmentally friendly operation with low energy consumption
  • Large, bright LED display with splash-proof keypad
  • Optional air or water cooling with easy filling and drain tap easily accessible
  • Circulating pumps with flow rates up to 60 L/min and 6 bar pressure