Condensators, Davies
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05215313 05215314 05215413 05215414
LENZ05215313EA 139 EUR
LENZ05215313 LENZ05215314 LENZ05215413 LENZ05215414
Condensators, Davies
Condensors
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two GL 14 threaded plastic olives.