Dimroth condensers for Soxleth extractors
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05246017 05246016 05246015 05246014 05246018
LENZ05246017EA 222 EUR
LENZ05246017 LENZ05246016 LENZ05246015 LENZ05246014 LENZ05246018
Dimroth condensers for Soxleth extractors
Condensors
DURAN® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Dimroth condensers for extractors, for condensing vapours. Water connection with two GL 14 threaded plastic olives.