Reageerbuisjes, met tuit
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6400160 6400165 6400166 6400170
HIRS6400160EA 9.2 EUR
HIRS6400160 HIRS6400165 HIRS6400166 HIRS6400170
Reageerbuisjes, met tuit
Buisjes Reageerbuisjes
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Graduated