Microtube racks, with lid, FLIPSRTIP™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8668 8678 8688 8698 8618 8608
700-2847EA 33.9 EUR
700-2847 700-2840 700-2841 700-2842 525-0032 MLBP8608
Microtube racks, with lid, FLIPSRTIP™
Rekken Tube Racks
Recycled plastic, two way rack.
  • Microplate format 96 holes (8×12)
  • Stable low profile design with alphanumeric grid
  • Protective lid
Holds 0,2 ml strips and 1,5 ml tubes on one side and 0,5 ml tubes on the other.