Droogbuizen, U-vormig
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05339010 05339013 05339018
LENZ05339010EA 24.4 EUR
LENZ05339010 LENZ05339013 LENZ05339018
Droogbuizen, U-vormig
Buisjes Droogbuizen
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).