Inoculatie-entogen met naalduiteinde, PS
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
631-2216EA 78.3 EUR
631-2216 631-2217 BRND452215
Inoculatie-entogen met naalduiteinde, PS
Uitstrijkspatels Entogen en -naalden
PS, gamma sterilised, disposable. For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface.