Laboratoriumflessen, smalle hals, met schroefdraad
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
704008 704010 704004 704006 704002 704020 704018 704016 704014 704012
215-3838EA 16.2 EUR
215-3838 215-3839 215-3836 215-3837 215-3835 215-3844 215-3843 215-3842 215-3841 215-3840
Laboratoriumflessen, smalle hals, met schroefdraad
Flessen Glazen flessen
Soda-lime glass, amber, coated or uncoated.
  • Good chemical and physical properties
  • Maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 °C
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably.