Trechters, PP
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7750101 7750102 7750103 7750104 7750105 7750106 7750107 7750108
HIRS7750101EA 34.6 EUR
HIRS7750101 HIRS7750102 HIRS7750103 HIRS7750104 HIRS7750105 HIRS7750106 HIRS7750107 HIRS7750108
Trechters, PP
Trechters Filtertrechters Filtertrechters
Funnels, made of PP, translucent. With smooth interior surfaces.
  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper