Poedertrechters
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7760101 7760102 7760104 7760105 7760103
HIRS7760101EA 28.5 EUR
HIRS7760101 HIRS7760102 HIRS7760104 HIRS7760105 HIRS7760103
Poedertrechters
Trechters Poedertrechters
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable