Wetzel Waterjet pumps
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05585000 05585200
LENZ05585000EA 76.8 EUR
LENZ05585000 LENZ05585200
Wetzel Waterjet pumps
Pompen Waterstraalpompen
Water jet pumps made of borosilicate glass with or without non-return valve.