Adapters, NS-kegel naar bolvormige verbindingshuls
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01521914 01521929 01522929 01523529
LENZ01521914EA 32.5 EUR
LENZ01521914 LENZ01521929 LENZ01522929 LENZ01523529
Adapters, NS-kegel naar bolvormige verbindingshuls
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters from ground joint cone to spherical joint socket.