Adapters, NS-kegel naar bolvormige verbindingskogel
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01531419 01532929 01532935 01532941
LENZ01531419EA 33.1 EUR
LENZ01531419 LENZ01532929 LENZ01532935 LENZ01532941
Adapters, NS-kegel naar bolvormige verbindingskogel
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.