2-Ethyl-1-butene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
E0067-500ML E0067-25ML E0067-100ML
TCIAE0067-500MLEA 909 EUR
TCIAE0067-500ML TCIAE0067-25ML TCIAE0067-100ML
2-Ethyl-1-butene ≥97.0% (by GC)
2-Ethyl-1-butene
Formule: C₆H₁₂
Kookpunt: 64°C
Dichtheid: 0,688 g/cm³ (24 °C)
Vlampunt: –10°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009341
CAS nummer: 760-21-4
EINECS: 212-078-5
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=566a3cba5316fd3ec8e969751444f007c64e08db&vid=8f11e725c65b007058ca828505cedb055e87f808&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %