Cyclohexylethene ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L10756.06
L10756.14
L10756.06EA
52.4
EUR
L10756.06
L10756.14
Cyclohexylethene ≥97%
Cyclohexylethene
Purity: 97%
CAS No.: 695-12-5
Molecular Formula: C8H14
Formula Weight: 110.20
Flash Point: 22°(71°F)
Refractive Index: 1.4465
Boiling Point: 127-128°
Beilstein Reference: 1304663
UN No.: 3295
MDL No.: MFCD00001514
Formule:
C₈H₁₄ Kookpunt: 127…128 °C Dichtheid: 0,805 Vlampunt: 22 °C (71 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001514 CAS nummer: 695-12-5 EINECS: 211-779-3 UN: 3295 ADR: 3,II |