3,3-Dimethyl-1-butyne 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
223440050 223440250
ACRO223440050EA 71.4 EUR
ACRO223440050 ACRO223440250
3,3-Dimethyl-1-butyne 98%
3,3-Dimethyl-1-butyne
Formule: C₆H₁₀
Kookpunt: 33…38 °C (760 mmHg)
Smeltpunt: –78 °C
Dichtheid: 0,66 g/cm³
Vlampunt: –034 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008852
CAS nummer: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3730 to 1.3750 (20°C, 589 nm)
GC ≥97.5 %