Centrifugeerbuizen, ASTM
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05675000 05675003 05675005
LENZ05675000EA 40.4 EUR
LENZ05675000 LENZ05675003 LENZ05675005
Centrifugeerbuizen, ASTM
Buisjes Centrifugebuisjes
DURAN®, borosilicate glass.
  • Withstands forces up to 700 ×g
  • Good temperature resistance
  • Good chemical resistance
With conical bottom or pear shaped.