Mengmaatcilinders, DURAN®, klasse A
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2340293 2340294 2340260 2340270 2340275 2340280 2340286 2340290
HIRS2340293EA 148 EUR
HIRS2340293 HIRS2340294 HIRS2340260 HIRS2340270 HIRS2340275 HIRS2340280 HIRS2340286 HIRS2340290
Mengmaatcilinders, DURAN®, klasse A
Cilinders Maatcilinders
DURAN®, borosilicate glass, with hollow glass stopper, class A, conformity certified.
  • Blue Graduation, main point graduation
  • Hexagonal glass base, NS ground socket
  • Conformity certified, with dated batch identification
Certificaten: DIN EN ISO 4788