Strips met 8 PCR-buisjes met losse doppenstrips
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
781326 781327
731-0208EA 254 EUR
731-0208 731-0209
Strips met 8 PCR-buisjes met losse doppenstrips
Buisjes PCR-buisjes
PP. Eight connected 0,2 ml tubes and separate domed or flat caps in strips of 8.
  • Flat caps are suitable for real-time PCR (qPCR)
  • Easy to open and close without tools
  • The tubes have a holding strap at one end, and the cover strips have a small lip on the side of each cap for careful, contamination-free opening
  • DNase-, RNase- and DNA-free
Colour: Clear