Stopcocks for burettes, lateral
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8901160 8901170 8901175
HIRS8901160EA 33.1 EUR
HIRS8901160 HIRS8901170 HIRS8901175
Stopcocks for burettes, lateral
Afsluitkranen
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
  • Excellent chemical resistance
  • Excellent temperature resistance