HyperSep™ glass block manifolds
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60104-232 60104-233
549-0597EA 1650 EUR
549-0597 549-0598
HyperSep™ glass block manifolds
Spruitstukken
Vacuum manifolds for HyperSep™ SPE columns allow easy processing of samples.
  • Vacuum chamber is a glass block allowing visible inspection of extraction process and easy cleaning
  • Vacuum control valve for optimizing sample flow rate
  • Rigid Corian® lid which resists warping with extended use
  • Bulkhead Luer fittings allow sample to elute directly into disposable test tubes
  • Lid has 16 or 24 positions in parallel rows and corner leg supports to prevent damage to luers and tips when lid is not in use
Leveringsinformatie: Supplied with glass block, Corian® manifold lid, cover gasket, vacuum gauge and valve assembly, 16 or 24 PTFE tips, adjustable collection rack, 16 or 24 bulkhead luer fittings and plugs.