PEEK tubing, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1559 1560 1560L 1560XL 1561 1561L 1561XL 1568 1568L 1568XL 1569 1569L 1531 1531B 1531BL 1531L 1532 1532L 1533 1533L 1533XL 1534 1535 1535L 1535XL 1536 1536L 1536XL 1538 1538L 1538XL 1569XL 1570 1571 1572 1574 1576 1576L 1576XL 1577 1577L 1577XL 1581 1581L 1581XL 1544
UPCH1559EA 116 EUR
UPCH1559 554-1818 UPCH1560L UPCH1560XL 554-1819 UPCH1561L UPCH1561XL UPCH1568 UPCH1568L UPCH1568XL 554-3284 UPCH1569L 554-1811 554-1812 UPCH1531BL UPCH1531L 554-1813 UPCH1532L 554-1814 UPCH1533L UPCH1533XL 554-1815 554-1816 UPCH1535L UPCH1535XL 554-1817 UPCH1536L UPCH1536XL 554-3283 UPCH1538L UPCH1538XL UPCH1569XL 554-3285 UPCH1571 UPCH1572 UPCH1574 UPCH1576 UPCH1576L UPCH1576XL UPCH1577 UPCH1577L UPCH1577XL UPCH1581 UPCH1581L UPCH1581XL UPCH1544
PEEK tubing, Upchurch Scientific®
Slangen Chromatography Tubing
Polymer tubing, coloured.
  • Biocompatible, inert and easy to cut
  • Great for high pressure applications
  • Many sizes colour-coded for easier inner diameter identification
PEEK (polyetheretherketone) tubing is biocompatible, chemically inert and can be used in place of stainless steel tubing in most liquid analytical systems. Additionally, when the outer diameter is smaller than 1/16”, PEEK tubing can be used in place of more traditional fused silica capillary tubing. PEEK tubing can be used with stainless steel nuts and ferrules, flangeless fittings, SuperMore Product Information