Prekolom- en inline filters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-316 C-420 A-100 A-101 A-102 A-224 A-315 A-315-01 A-315-02 A-318
554-1783EA 172 EUR
554-1783 UPCHC-420 UPCHA-100 UPCHA-101 UPCHA-102 554-2204 554-1782 554-3020 554-3021 554-1784
Prekolom- en inline filters, Upchurch Scientific®
Filters Voorkolom filters en frits
These filters are ideal for higher pressure applications and to help prevent particulate contamination.
  • For ¹/₁₆" Ø tubing