QuEChERS extraction products
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60105-215 60105-208 60105-230 60105-333 60105-332
549-0766EA 0 EUR
549-0766 549-0764 HYPE60105-230 HYPE60105-333 HYPE60105-332
QuEChERS extraction products
Kolommen voor chromatografie
The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for the extraction of pesticide residues in fruit, vegetables and other foods. The QuEChERS method offers the advantage of high recoveries, accurate results and high sample throughput, combined with the associated cost savings with low solvent and labour requirements. Yet the procedure is robust and rugged.
  • Simple manual procedure
  • Determine wide range of pesticide types
  • Less solvent usage than conventional techniques
  • High recoveries, accurate results and high sample throughput