Gasmonsterzakjes, Chemware®
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D1075005 D1075001 D1075004 D1075006
300-0275EA 70.2 EUR
300-0275 300-0271 300-0274 300-0276
Gasmonsterzakjes, Chemware®
Zakken Gaszakken
These flexible, sturdy bags, made from 5 mil thick FEP fluoropolymer film, are heatsealed on all four sides for complete chemical resistance and leakproof operation. Can be used for gas sample analysis, extracting air samples from smokestack exhaust systems, monitoring air quality or calibrating instruments in the field.
  • Easy to use
  • Flexible and sturdy
  • Completely chemical-resistant
  • Wil not contaminate samples
  • Leak proof, heat sealed all four sides
  • Completely chemical-resistant
  • Low permeability – no sample loss
  • No aging – unlimited storage life
  • Unique seam construction for added strength
The bags are available with either a nickel-plated, brass on/off valve that accepts ¹/₄" Int.Ø flexible tubing or with PTFE/silicone septum in polypropylene housing for use with a hypodermic needle. Because of the precision of manufacturing controls, Chemware® gas sampling bags do not require a second outboard seal. The single seam resists most corrosives and is suitable for use where difficult chemical conditions exist.
Bestelinformatie: Individually packaging. Custom sizes and configurations available upon request.