Micro capillaries, disposable, Openend®
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
905022001 905021001
HIRS905022001EA 141 EUR
HIRS905022001 HIRS905021001
Micro capillaries, disposable, Openend®
Pipetten Micro-capillairen
Openend®, disposable capillary pipettes, sodium heparinized, DURAN®, certified conformity, with predefined breaking point for exact volume configuration.
  • Openend is filled by capillary action beyond the middle to approximately the printed ring mark
  • Residual medium on the outside of the capillary is removed
  • Capillary separated through gentle pressure with the fingers
  • The completely filled part now contains the exact volume