2-Bromobenzaldehyde 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: o-Bromobenzaldehyde

106650100 106650250 106651000
ACRO106650100EA 68.7 EUR
ACRO106650100 ACRO106650250 ACRO106651000
2-Bromobenzaldehyde 97%
2-Bromobenzaldehyde
Formule: C₇H₅BrO
Kookpunt: 230 °C (760 mmHg)
Smeltpunt: 21…22 °C
Dichtheid: 1,58 g/cm³
Vlampunt: 95 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003300
CAS nummer: 6630-33-7
EINECS: 229-622-2

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5950 to 1.5970 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR