4-Bromobenzaldehyde for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.04146.0010 8.04146.0100
8.04146.0010EA 56.8 EUR
8.04146.0010 8.04146.0100
4-Bromobenzaldehyde for synthesis, Sigma-Aldrich®
4-Bromobenzaldehyde
Formule: C₇H₅BrO
Kookpunt: 244 °C (1013 hPa)
Smeltpunt: 54…56 °C
Dichtheid: 1,849 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003377
CAS nummer: 1122-91-4
EINECS: 214-365-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 55 - 58 °C
Molar Mass 185.02 g/mol
Bulk Density 670 kg/m3
Boiling Point 123 - 125 °C (31 hPa)
Flash Point 108 °C
Density 1.85 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR